Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080969.3 | NP_001074438.1 | 1 - 2 | 227 | 114 | |
GenBank mRNA | AK010876.1 | - | 1 - 2 | 219 | 114 | |
BC092541.1 | - | 1 - 2 | 207 | 30280958 | 114 | |
BC115560.1 | - | 1 - 2 | 215 | 40101901 | 114 | |
BC115561.1 | - | 1 - 2 | 40101902 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |