Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027941.1 | NP_082217.1 | 6 - 7 | 667 | 69 | |
GenBank mRNA | AK005720.1 | - | 6 - 7 | 719 | 69 | |
BC100519.1 | - | 6 - 7 | 667 | 30919449 | 69 | |
BC116419.1 | - | 5 - 6 | 500 | 69 | ||
BC116420.1 | - | 5 - 6 | 507 | 40103399 | 69 | |
JF934738.1 | - | 6 - 7 | 618 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |