Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040395.4 | NP_001035485.2 | 6 - 7 | 990 | 76 | |
NM_001085410.2 | NP_001078879.1 | 6 - 7 | 990 | 76 | ||
NM_001286253.1 | NP_001273182.1 | 6 - 7 | 990 | 76 | ||
NM_001286255.1 | NP_001273184.1 | 5 - 6 | 906 | 76 | ||
GenBank mRNA | AK003858.1 | - | 6 - 7 | 567 | 76 | |
AK016988.1 | - | 6 - 7 | 556 | 76 | ||
AK046666.1 | - | 6 - 7 | 991 | 76 | ||
AK078318.1 | - | 6 - 7 | 593 | 76 | ||
AK149267.1 | - | 5 - 6 | 486 | 76 | ||
AK162601.1 | - | 6 - 7 | 564 | 76 | ||
BC115776.1 | - | 6 - 7 | 569 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |