Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025609.2 | NP_079885.2 | 1 - 2 | 40 | 80 | |
GenBank mRNA | AB088136.1 | - | 1 - 2 | 40 | 80 | |
BC027054.1 | - | 1 - 2 | 32 | 80 | ||
BC041110.1 | - | 1 - 2 | 35 | 80 | ||
BC054369.1 | - | 1 - 2 | 40 | 5356886 | 80 | |
BC114637.1 | - | 1 - 2 | 61 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |