Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024169.3 | NP_077131.2 | 1 - 2 | 238 | 59 | |
GenBank mRNA | AK003331.1 | - | 1 - 2 | 239 | 59 | |
AK007813.1 | - | 1 - 2 | 202 | 59 | ||
BC002311.1 | - | 1 - 2 | 192 | 59 | ||
BC022900.1 | - | 1 - 2 | 192 | 59 | ||
BC037596.1 | - | 1 - 2 | 194 | 5250344 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |