Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021790.2 | NP_068562.1 | 1 - 2 | 265 | 142 | |
GenBank mRNA | AB043687.1 | - | 1 - 2 | 265 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |