Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139299.2 | NP_647460.2 | 12 - 13 | 1634 | 74 | |
GenBank mRNA | AB083111.1 | - | 12 - 13 | 1633 | 74 | |
AF486621.1 | - | 9 - 10 | 1213 | 74 | ||
AY509150.1 | - | 1530 | 74 | |||
AY509151.1 | - | 1530 | 74 | |||
BC066164.1 | - | 10 - 11 | 1355 | 30475340 | 74 | |
BC145084.1 | - | 11 - 12 | 1537 | 74 | ||
BC145085.1 | - | 12 - 13 | 1660 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |