Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028015.2 | NP_082291.1 | 1 - 2 | 332 | 108 | |
GenBank mRNA | AK010241.1 | - | 1 - 2 | 260 | 108 | |
AK075694.1 | - | 1 - 2 | 252 | 108 | ||
AK145589.1 | - | 1 - 2 | 240 | 108 | ||
AK145762.1 | - | 1 - 2 | 254 | 108 | ||
AK146220.1 | - | 1 - 2 | 254 | 108 | ||
AK146417.1 | - | 1 - 2 | 223 | 108 | ||
AK219835.1 | - | 1 - 2 | 42 | 108 | ||
AY029533.1 | - | 1 - 2 | 252 | 108 | ||
BC043059.1 | - | 279 | 6419704 | 108 | ||
BC046797.1 | - | 1 - 2 | 332 | 6403129 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |