Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016518.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016518.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016518.mm.1 |
TC1500002350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016520.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016520.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016520.mm.1 |
TC1500002350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016521.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016521.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016521.mm.1 |
TC1500002350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016519.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016519.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016519.mm.1 |
TC1500002350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016517.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016517.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016517.mm.1 |
TC1500002350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016522.mm.1 |
NM_009948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016522.mm.1 |
NM_009948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016522.mm.1 |
TC1500002350.mm.1 |
MTA TC |