Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016704.2 | NP_057913.2 | 14 - 15 | 2231 | 105 | |
GenBank mRNA | AF184900.1 | - | 14 - 15 | 2232 | 105 | |
BC011251.1 | - | 14 - 15 | 2220 | 4196330 | 105 | |
BX515704.1 | - | 2 - 3 | 215 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |