Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021336.4 | NP_067311.4 | 3 - 4 | 155 | ||
GenBank mRNA | AF230356.1 | - | 3 - 4 | 155 | ||
AK005256.1 | - | 3 - 4 | 155 | |||
AK088438.1 | - | 3 - 4 | 155 | |||
AK151800.1 | - | 3 - 4 | 155 | |||
AK152546.1 | - | 3 - 4 | 155 | |||
AK153502.1 | - | 3 - 4 | 155 | |||
AK206837.1 | - | 2 - 3 | 155 | |||
BC029642.1 | - | 3 - 4 | 3588138 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |