Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
NM_001038696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
NM_001038696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
NM_001287015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
NM_026043.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
NM_026043.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021982.mm.1 |
TC0300003248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
NM_001038696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
NM_001038696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
NM_001287015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
NM_026043.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
NM_026043.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021978.mm.1 |
TC0300003248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
NM_001038696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
NM_001038696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
NM_001287015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
NM_026043.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
NM_026043.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021979.mm.1 |
TC0300003248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
NM_001038696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
NM_001038696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
NM_001287015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
NM_026043.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
NM_026043.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021980.mm.1 |
TC0300003248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
NM_001038696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
NM_001038696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
NM_001287015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
NM_026043.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
NM_026043.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021981.mm.1 |
TC0300003248.mm.1 |
MTA TC |