Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026796.1 | NP_081072.1 | 2 - 3 | 243 | 74 | |
GenBank mRNA | AK145261.1 | - | 2 - 3 | 248 | 74 | |
AK150857.1 | - | 2 - 3 | 247 | 74 | ||
AK164053.1 | - | 2 - 3 | 209 | 74 | ||
BC023119.1 | - | 2 - 3 | 243 | 4458755 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |