Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159850.1 | NP_001153322.1 | 4 - 5 | 901 | 87 | |
NM_001281847.1 | NP_001268776.1 | 4 - 5 | 792 | 87 | ||
NM_001281848.1 | NP_001268777.1 | 3 - 4 | 633 | 87 | ||
NM_010607.3 | NP_034737.2 | 4 - 5 | 997 | 87 | ||
GenBank mRNA | AK138427.1 | - | 4 - 5 | 673 | 87 | |
AK140716.1 | - | 4 - 5 | 672 | 87 | ||
AK164197.1 | - | 3 - 4 | 633 | 87 | ||
AY736359.1 | - | 4 - 5 | 636 | 87 | ||
BC062094.1 | - | 4 - 5 | 615 | 6848897 | 87 | |
U73488.2 | - | 5 - 6 | 1074 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |