Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141981.1 | NP_001135453.1 | 3 - 4 | 487 | 112 | |
NM_001141982.1 | NP_001135454.1 | 3 - 4 | 406 | 112 | ||
NM_030243.4 | NP_084519.1 | 3 - 4 | 380 | 112 | ||
GenBank mRNA | AK002739.1 | - | 3 - 4 | 534 | 112 | |
BC003333.1 | - | 3 - 4 | 364 | 3256678 | 112 | |
BU697339.1 | - | 3 - 4 | 406 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |