Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109761.2 | NP_001103231.1 | 14 - 15 | 1732 | 74 | |
NM_001177799.1 | NP_001171270.1 | 14 - 15 | 1999 | 74 | ||
NM_007601.3 | NP_031627.2 | 17 - 18 | 2275 | 74 | ||
GenBank mRNA | AF091998.1 | - | 14 - 15 | 1696 | 74 | |
AF127766.2 | - | 14 - 15 | 1719 | 74 | ||
AK049994.1 | - | 4 - 5 | 161 | 74 | ||
AK088613.1 | - | 2 - 3 | 486 | 74 | ||
BC090661.1 | - | 14 - 15 | 1732 | 30681055 | 74 | |
BC139790.1 | - | 17 - 18 | 1995 | 40047690 | 74 | |
X92523.1 | - | 17 - 18 | 1995 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |