Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007947.3 | NP_031973.2 | 5 - 6 | 567 | 132 | |
GenBank mRNA | U68546.1 | - | 5 - 6 | 463 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |