Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177600.4 | NP_808268.2 | 2 - 3 | 327 | 132 | |
GenBank mRNA | BB615231.1 | - | 2 - 3 | 327 | 130 | |
BC095976.1 | - | 1 - 2 | 73 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |