Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147778.3 | NP_680087.1 | 5 - 6 | 412 | 71 | |
GenBank mRNA | AK007318.1 | - | 5 - 6 | 417 | 71 | |
AK041634.1 | - | 5 - 6 | 413 | 71 | ||
AK145955.1 | - | 5 - 6 | 440 | 71 | ||
AK145999.1 | - | 5 - 6 | 442 | 71 | ||
BC038077.1 | - | 5 - 6 | 454 | 4013160 | 71 | |
BC048815.1 | - | 5 - 6 | 423 | 3482368 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |