Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026944.1 | NP_081220.1 | 2 - 3 | 201 | 54 | |
GenBank mRNA | AK145586.1 | - | 2 - 3 | 587 | 54 | |
AK187483.1 | - | 2 - 3 | 210 | 54 | ||
BC018196.1 | - | 2 - 3 | 201 | 3993214 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |