Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026752.4 | NP_081028.3 | 5 - 6 | 596 | 98 | |
GenBank mRNA | AK003661.1 | - | 5 - 6 | 493 | 98 | |
AK155300.1 | - | 5 - 6 | 597 | 98 | ||
AK171099.1 | - | 5 - 6 | 537 | 98 | ||
AK192012.1 | - | 3 - 4 | 206 | 98 | ||
BC019521.1 | - | 5 - 6 | 526 | 4220205 | 98 | |
BC132248.1 | - | 5 - 6 | 496 | 40130525 | 98 | |
BC132250.1 | - | 5 - 6 | 496 | 40130527 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |