Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145376.5 | NP_663351.3 | 1 - 2 | 135 | 65 | |
GenBank mRNA | AB244717.2 | - | 1 - 2 | 135 | 65 | |
AK081865.1 | - | 1 - 2 | 293 | 65 | ||
AK132379.1 | - | 1 - 2 | 269 | 66 | ||
AK155286.1 | - | 1 - 2 | 299 | 65 | ||
AK169190.1 | - | 1 - 2 | 268 | 65 | ||
AK170723.1 | - | 1 - 2 | 288 | 65 | ||
AK171582.1 | - | 1 - 2 | 288 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |