Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145780.1 | NP_001139252.1 | 3 - 4 | 408 | 78 | |
NM_025917.4 | NP_080193.1 | 2 - 3 | 195 | 78 | ||
GenBank mRNA | AF353245.1 | - | 2 - 3 | 188 | 78 | |
AK003257.1 | - | 2 - 3 | 188 | 78 | ||
AK003385.1 | - | 1 - 2 | 59 | 78 | ||
AK008573.1 | - | 2 - 3 | 188 | 78 | ||
AK008661.1 | - | 1 - 2 | 93 | 78 | ||
AK012490.1 | - | 2 - 3 | 189 | 78 | ||
AK160726.1 | - | 2 - 3 | 188 | 78 | ||
AK165639.1 | - | 2 - 3 | 337 | 78 | ||
AY842453.1 | - | 2 - 3 | 174 | 78 | ||
BC075695.1 | - | 3 - 4 | 314 | 30623689 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |