Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178720.4 | NP_848835.1 | 7 - 8 | 842 | 130 | |
GenBank mRNA | AK034628.1 | - | 7 - 8 | 843 | 130 | |
AK047981.1 | - | 6 - 7 | 752 | 130 | ||
AK052599.1 | - | 8 - 9 | 952 | 130 | ||
BC116254.1 | - | 7 - 8 | 811 | 40093631 | 130 | |
BC116255.1 | - | 7 - 8 | 811 | 40093632 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |