Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024142.1 | NP_001019313.1 | 4 - 5 | 1675 | 65 | |
GenBank mRNA | AK142389.1 | - | 4 - 5 | 1670 | 65 | |
BC052485.1 | - | 3 - 4 | 1142 | 65 | ||
BC054731.1 | - | 4 - 5 | 1665 | 65 | ||
BC062126.1 | - | 3 - 4 | 1034 | 65 | ||
BC094565.1 | - | 3 - 4 | 1459 | 6836944 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |