Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027307.4 | NP_081583.3 | 2 - 3 | 196 | 72 | |
GenBank mRNA | BC098486.1 | - | 2 - 3 | 192 | 30024953 | 72 |
CD552063.1 | - | 2 - 3 | 157 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |