Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136260.1 | NP_001129732.1 | 21 - 22 | 2825 | 85 | |
NM_001197147.1 | NP_001184076.1 | 21 - 22 | 2798 | 85 | ||
NM_018760.2 | NP_061230.2 | 21 - 22 | 2825 | 85 | ||
GenBank mRNA | AW124701.1 | - | 2 - 3 | 85 | ||
BC026592.1 | - | 12 - 13 | 1618 | 85 | ||
BC129863.1 | - | 21 - 22 | 2787 | 85 | ||
BC129864.1 | - | 21 - 22 | 2786 | 85 | ||
BC148292.2 | - | 21 - 22 | 2761 | 40110074 | 85 | |
CB524421.1 | - | 6 - 7 | 686 | 85 | ||
HQ018820.1 | - | 18 - 19 | 2662 | 85 | ||
HQ204220.1 | - | 21 - 22 | 2769 | 85 | ||
HQ285250.1 | - | 18 - 19 | 2638 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |