Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012007.1 | NP_036137.1 | 6 - 7 | 858 | 75 | |
GenBank mRNA | AF136272.1 | - | 6 - 7 | 858 | 75 | |
AF158182.1 | - | 5 - 6 | 798 | 75 | ||
AK005526.1 | - | 6 - 7 | 865 | 75 | ||
AK131661.1 | - | 6 - 7 | 865 | 75 | ||
BC103769.1 | - | 6 - 7 | 854 | 30787117 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |