Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183029.2 | NP_898850.2 | 4 - 5 | 676 | 74 | |
GenBank mRNA | AK044984.1 | - | 3 - 4 | 192 | 74 | |
AK049196.1 | - | 4 - 5 | 373 | 74 | ||
AK170531.1 | - | 3 - 4 | 180 | 74 | ||
AY531659.1 | - | 4 - 5 | 340 | 74 | ||
BC023758.1 | - | 4 - 5 | 374 | 74 | ||
BC054552.1 | - | 4 - 5 | 396 | 74 | ||
CB290059.1 | - | 4 - 5 | 676 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |