Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163233.1 | NP_001156705.1 | 12 - 13 | 1829 | 67 | |
NM_053081.2 | NP_444311.1 | 12 - 13 | 1850 | 67 | ||
GenBank mRNA | AF112439.1 | - | 12 - 13 | 1659 | 67 | |
AY049715.1 | - | 12 - 13 | 1650 | 67 | ||
BC050890.1 | - | 12 - 13 | 2107 | 67 | ||
BC139763.1 | - | 12 - 13 | 1632 | 40110334 | 67 | |
BC141282.1 | - | 12 - 13 | 1666 | 9055894 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |