Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081141.1 | NP_001074610.1 | 10 - 11 | 1809 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Mus musculus Inventoried Probe spans exons |