Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290430.1 | NP_001277359.1 | 10 - 11 | 1026 | 70 | |
NM_133991.2 | NP_598752.2 | 10 - 11 | 1032 | 70 | ||
GenBank mRNA | AK036313.1 | - | 10 - 11 | 990 | 70 | |
AK150251.1 | - | 7 - 8 | 1120 | 70 | ||
BC034880.1 | - | 10 - 11 | 896 | 3468633 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |