Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008228.2 | NP_032254.1 | 9 - 10 | 998 | 74 | |
GenBank mRNA | BC092070.1 | - | 9 - 10 | 4217199 | 74 | |
BC092285.1 | - | 9 - 10 | 74 | |||
BC108371.1 | - | 9 - 10 | 4976514 | 74 | ||
U80780.1 | - | 9 - 10 | 74 | |||
X98207.1 | - | 9 - 10 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |