Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190325.1 | NP_001177254.1 | 1 - 2 | 291 | 151 | |
GenBank mRNA | AK137514.1 | - | 1 - 2 | 292 | 151 | |
AK137552.1 | - | 1 - 2 | 292 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |