Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028384.1 | NP_082660.1 | 5 - 6 | 659 | 63 | |
GenBank mRNA | AK158659.1 | - | 3 - 4 | 439 | 63 | |
BC147354.1 | - | 3 - 4 | 404 | 9056407 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |