Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081633.1 | NP_001075102.1 | 8 - 9 | 1087 | 127 | |
NM_022408.2 | NP_071853.2 | 8 - 9 | 1090 | 127 | ||
GenBank mRNA | AF037256.1 | - | 8 - 9 | 1089 | 127 | |
AK088520.1 | - | 8 - 9 | 1052 | 127 | ||
AK142684.1 | - | 8 - 9 | 1055 | 127 | ||
AK148404.1 | - | 8 - 9 | 1058 | 127 | ||
BC013711.1 | - | 8 - 9 | 1066 | 3708646 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |