Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snora15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003681.1 | 1 | 63 | 85 | ||
GenBank mRNA | AF357410.1 | - | 1 | 63 | 85 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream |
chaperonin containing Tcp1, subunit 6a (zeta) |
Cct6a |
CCT-zeta, CCT-zeta-1, Cct6, Cctz-1, TCP-1-zeta |
50.3kb | Sense | Chr. 5 - 129844962 - 129846443 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |