Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034868.3 | NP_001030040.2 | 1 - 2 | 120 | 100 | |
GenBank mRNA | AK148121.1 | - | 1 - 2 | 51 | 100 | |
BC141225.1 | - | 1 - 2 | 43 | 9055837 | 100 | |
CJ131827.1 | - | 1 - 2 | 171 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |