Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080434.3 | NP_536682.2 | 2 - 3 | 170 | 73 | |
GenBank mRNA | AF327059.1 | - | 2 - 3 | 170 | 73 | |
AK004903.1 | - | 2 - 3 | 173 | 73 | ||
AK004936.1 | - | 2 - 3 | 170 | 73 | ||
AK050280.1 | - | 2 - 3 | 171 | 73 | ||
BC011198.1 | - | 3 - 4 | 196 | 4196296 | 73 | |
BC029334.1 | - | 2 - 3 | 185 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |