Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021518.3 | NP_067493.1 | 1 | 151 | 94 | |
GenBank mRNA | AK042862.1 | - | 1 | 121 | 94 | |
AK079071.1 | - | 1 | 152 | 94 | ||
AK135421.1 | - | 1 | 135 | 94 | ||
AK146179.1 | - | 1 | 136 | 94 | ||
AK150030.1 | - | 1 | 183 | 94 | ||
AK159607.1 | - | 1 | 172 | 94 | ||
AK160333.1 | - | 1 | 94 | |||
AK163320.1 | - | 1 | 134 | 94 | ||
BC058382.1 | - | 1 | 149 | 6820798 | 94 | |
X95403.1 | - | 1 | 139 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |