Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104435.1 | 10 - 11 | 2891 | 84 | ||
NR_104436.1 | 10 - 11 | 2891 | 84 | |||
NR_104437.1 | 10 - 11 | 2891 | 84 | |||
NR_104438.1 | 10 - 11 | 2891 | 84 | |||
GenBank mRNA | AB059645.1 | - | 10 - 11 | 2884 | 84 | |
AB059646.1 | - | 10 - 11 | 2884 | 84 | ||
AB059647.1 | - | 10 - 11 | 2884 | 84 | ||
AB059648.1 | - | 10 - 11 | 2884 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
dicer 1, ribonuclease type III |
Dicer1 |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
-23.9kb | Sense | Chr. 12 - 104687740 - 104751952 on NCBI Build GRCm38 |
Downstream |
spectrin repeat containing, nuclear envelope family member 3 |
Syne3 | nesprin-3 | 152.7kb | Sense | Chr. 12 - 104929830 - 105010098 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |