Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975426.1 | XP_027831227.1 | 1 - 2 | 824 | 64 | |
XM_027975427.1 | XP_027831228.1 | 1 - 2 | 824 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |