Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131031.1 | NP_001124503.1 | 4 - 5 | 571 | 88 | |
GenBank mRNA | EU797608.1 | - | 4 - 5 | 571 | 88 |