Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022306.4 | XP_004022355.1 | 4 - 5 | 991 | 67 | |
XM_027962875.1 | XP_027818676.1 | 6 - 7 | 1106 | 67 | ||
XM_027962876.1 | XP_027818677.1 | 5 - 6 | 938 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |