Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977542.1 | XP_027833343.1 | 1 - 2 | 75 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |