Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163052.1 | NP_001156524.1 | 5 - 6 | 841 | 99 | |
XM_012183366.3 | XP_012038756.1 | 6 - 7 | 949 | 99 | ||
GenBank mRNA | FJ958348.1 | - | 5 - 6 | 841 | 99 |