Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199068.1 | NP_001185997.1 | 5 - 6 | 682 | 101 | |
GenBank mRNA | HQ433587.1 | - | 5 - 6 | 682 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |