Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975258.1 | XP_027831059.1 | 4 - 5 | 891 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |