Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003968.4 | XP_004004017.2 | 9 - 10 | 1274 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |