Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103585.3 | XP_011958975.2 | 2 - 3 | 391 | 99 | |
XM_012103586.3 | XP_011958976.2 | 2 - 3 | 723 | 99 |